Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucralfate, Fine Powder, USP, 30-38%, Spectrum™ Chemical
SDP

Catalog No. 18607065 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-065 50 kg Fiber Drum
18-607-067 500 g Amber Glass Bottle
18-607-066 5 kg Poly Pail
18-607-068 25 kg Fiber Drum
18-607-063 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-065 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S176450KGBL
Add to Cart
Add to Cart
Request a Quote

S1764, 54182-58-0, C12H54Al16O75S8

Spectrum™ Chemical Sucralfate, Fine Powder, USP is an oral gastrointestinal medicine used in the treatment of duodenal ulcers. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 54182-58-0

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 50 kg
Percent Purity 30 to 38%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.