Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucralose 98.0+%, TCI America™
SDP

Catalog No. p-7135956 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08395G 5 g
S083925G 25 g
S0839100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S08395G Supplier TCI America Supplier No. S08395G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56038-13-2
Molecular Formula C12H19Cl3O8
Molecular Weight (g/mol) 397.63
MDL Number MFCD03648615
InChI Key BAQAVOSOZGMPRM-QBMZZYIRSA-N
Synonym sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvit
PubChem CID 71485
ChEBI CHEBI:32159
IUPAC Name (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol
SMILES OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl

Specifications

Melting Point 130°C
Color White
Quantity 5 g
Formula Weight 397.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sucralose
RTECSNumber LW5440140
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.