Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sucrose
SCI_ED

Catalog No. S25590B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Reagent Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25590B 2.5 kg Reagent Grade
S25590 500 g Reagent Grade
S25590A 1 kg Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25590B
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade Sucrose suitable for educational use and lab testing.

  • Sucrose
  • Reagent – suitable for educational applications
  • Quantity 2.5 kg
  • Density approx. 1.6 g/mL
  • Reagent grade container
  • Linear formula C12H22O11
  • Formula weight 342.3 g/mol
  • Physical form solid (powder)
  • Certificate of Analysis (C of A) not available

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Color White
Density ∽1.6g/mL
Packaging Reagent Grade
Linear Formula C12H22O11
Quantity 2.5 kg
Formula Weight 342.3
Grade Reagent
Chemical Name or Material Sucrose
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.