Learn More
Sucrose, 99% max., Molecular biology reagent grade, MP Biomedicals™
Shop All MP Biomedicals, Inc ProductsSpecifications
Specifications
| Color | White |
| Density | 1.587g/cm3 at 25°C |
| Assay Percent Range | >99% |
| Quantity | 500 g |
| Formula Weight | 342.3 |
| Percent Purity | ≥99% |
| Grade | Molecular Biology Reagent |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sucrose |
Safety and Handling
| EINECSNumber | 200-334-9 |
| Recommended Storage | Store at room temperature. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.