Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sucrose, ACS

Catalog No. AA3650830
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3650830 250 g
AA36508A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3650830 Supplier Thermo Scientific Chemicals Supplier No. 03650830
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Edge

missing translation for 'validMainframeMsg'

CAS: 57-50-1 | C12H22O11 | 342.30 g/mol

Sucrose is widely utilized in medications to carry a more pleasant taste to indigestible chemicals. It is used in many medical dosage forms such as chewable tablets, syrups, tablets, or glues. Sucrose is mainly used in foods nowadays. The fatty acid esters of sucrose show useful application in transdermal therapeutic systems. Sucrose-gradient centrifugation also plays an important role in the selective isolation of nocardia spp. from soil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 185°C to 187°C
Density 1.58 g/mL
Quantity 250 g
Beilstein 90825
Merck Index 14,8881
Solubility Information Soluble in water; Slightly soluble in alcohol,methanol; Insoluble in diethyl ether.
Formula Weight 342.3
Grade ACS
Physical Form Crystalline
Chemical Name or Material Sucrose, For analysis ACS
EINECSNumber 200-334-9
RTECSNumber WN6500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.