Learn More
Roche Diagnostics Sucrose Bio HT, 200 tests
Supplier:  Roche Diagnostics 07705565001
Encompass
For control of culture media and for monitoring in a running culture process, the sucrose concentration can be determined on a Cedex Bio or Cedex Bio HT Analyzer with high reliability and convenience.
Catalog No. NC1285674
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.