Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, Crystal, NF, Spectrum™ Chemical
SDP

Catalog No. 18607095 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-095 45 kg Poly Bottle
18-607-092 2.5 kg Poly Bottle
18-607-091 6 x 500 g Poly Bottle
18-607-093 4 x 2.5 kg Poly Bottle
18-607-094 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-095 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SU10345KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SU103, 57-50-1, C12H22O11

Spectrum™ Chemical Sucrose, Crystal, NF is used in medications to impart a more pleasant taste to often unpalatable chemicals and can be found in many medical dosage forms including chewable tablets, syrups, lozenges, or gums. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
Loss on Drying 0.001
Packaging Poly Bottle
Quantity 45 kg
Specific Rotation +66.3° to +67.0°
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sucrose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.