Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| Acidity | Pass Test (BP/EP) |
| Melting Point | 190°C |
| Color | White |
| pH | 6.5 to 7.5 |
| Ignition Residue | 0.05% max. (NF) |
| Loss on Drying | 0.1% max. (BP/EP) |
| Packaging | Poly Pail |
| Quantity | 12 kg |
| Specific Rotation | +66.3 to +67.0° (+ or -) (BP/EP), 65.9° min. (NF) |
| Residual Solvents | Meets Requirements (BP/EP/NF) |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. NFPA Health:1 Flammability:1 Instability:0 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?