Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sucrose, Molecular Biology Grade

Catalog No. AAJ65148A1
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65148A1 1 kg
AAJ6514836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65148A1 Supplier Thermo Scientific Chemicals Supplier No. J65148A1
Add to Cart

missing translation for 'validMainframeMsg'

For protein purification by sucrose density gradients | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol

Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 185°C to 187°C
Color White
Density 1.58
Odor Odorless
Quantity 1 kg
Beilstein 90825
Solubility Information Soluble in water
Formula Weight 342.3
Grade Molecular Biology
Physical Form Crystalline Solid
Chemical Name or Material Sucrose
Show More Show Less
EINECSNumber 200-334-9
RTECSNumber WN6500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.