Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, MP Biomedicals™

Catalog No. mp021902715 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021902715 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021902715 Supplier MP Biomedicals, Inc Supplier No. 02190271.5
Only null left
Add to Cart
Add to Cart

Used for a variety of applications. Sucrose, MP Biomedicals is a simple carbohydrate isolated from cane sugar; it is not synthetic. Sucrose is a disaccharide composed of glucose and fructose that can be used to prepare density gradients, culture media and

Sucrose can be used to prepare density gradients for cell/organelle separation. The refractive index of the solution will be directly related to the density. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Absorbance A50%/260 ≤0.15; A50%/280 ≤0.15 (Lit.)
Melting Point 177°C to 187°C
Density 1.587 at 25°C (water=1) (Lit.)
Quantity 500 g
Solubility Information Soluble in water (1g/0.5mL at 25°C; 1g/0.2mL boiling water), ethanol (1g/170mL) or methanol (1g/100mL); moderately soluble in glycerol or pyridine.
Optical Rotation 66.0° ± 5 (H2O, NH4OH)
Formula Weight 342.30
Percent Purity ≥95%
Physical Form Crystalline Powder
Recommended Storage Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.