Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose Octaacetate, NF, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607079 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-079 500 g Poly Bottle
18-607-078 125 g Poly Bottle
18-607-081 12 kg Poly Bottle
18-607-083 25 kg Poly Bottle
18-607-084 50 kg Poly Bottle
18-607-082 25 g Poly Bottle
18-607-080 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-079 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SU104500GM
Add to Cart
Add to Cart

SU104, 126-14-7, C28H38O19

Spectrum™ Chemical Sucrose Octaacetate, NF is used as a bitterant and aversive agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 126-14-7
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.59
InChI Key ZIJKGAXBCRWEOL-SAXBRCJISA-N
IUPAC Name [(2R,3R,4S,5S)-3,4-bis(acetyloxy)-5-[(acetyloxy)methyl]-5-{[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxolan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@@](COC(C)=O)(O[C@H]2O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]2OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

Specifications

CAS Max % 1
Boiling Point 250°C (482°F)
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98 to 100.5%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sucrose Octaacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.