Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, Ultrapure, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18607097 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
6 x 1 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-097 1 kg Poly Bottle
18-607-096 500 g Poly Bottle
18-607-099 5 kg Poly Pail
18-607-098 6 x 1 kg Poly Bottle
18-607-100 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-097 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S16971KG
Only null left
Add to Cart
Add to Cart

S1697, 57-50-1, C12H22O11

Spectrum™ Chemical Sucrose, Ultrapure , also known as sugar, is derived from cane sugar and used as a food sweetener. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 1 kg
Specific Rotation +66.3° to +66.8°
Percent Purity 99.9%
Grade Ultrapure
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.