Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan II 93.0+%, TCI America™
SDP

Catalog No. O010125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O010125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O010125G Supplier TCI America Supplier No. O010125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3118-97-6
Molecular Formula C18H16N2O
Molecular Weight (g/mol) 276.34
MDL Number MFCD00003896
InChI Key YPXOPAFVVHXQDP-ZZEZOPTASA-N
Synonym sudan ii, solvent orange 7, sudan red, oil red xo, fettorange b, pyronalrot r, sudan orange, sudan ax, c.i. solvent orange 7, motirot g
PubChem CID 5809936
IUPAC Name (1Z)-1-[2-(2,4-dimethylphenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES CC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(C)=C1

Specifications

Melting Point 160°C
Quantity 25 g
Formula Weight 276.34
Percent Purity ≥93.0% (E)
Physical Form Crystalline Powder
Chemical Name or Material Sudan II [=1-(2,4-Xylidylazo)-2-naphthol]
EINECSNumber (5)-3068
RTECSNumber QL5850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.