accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNOCH3(Z)

CAS RN 3118-97-6

IUPAC Name: (1Z)-1-[2-(2,4-dimethylphenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Synonyms: colouring agent CI 12140
Molecular Weight (g/mol): 276.34
Molecular Formula: C18H16N2O
InChi Key: YPXOPAFVVHXQDP-ZZEZOPTASA-N
SMILES: CC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(C)=C1
Grade 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)