Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sudan II, pure

Catalog No. AC190150250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC190150250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC190150250 Supplier Thermo Scientific Chemicals Supplier No. 190150250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3118-97-6 | C18H16N2O | 276.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3118-97-6
Molecular Formula C18H16N2O
Molecular Weight (g/mol) 276.34
MDL Number MFCD00003896
InChI Key YPXOPAFVVHXQDP-ZZEZOPTASA-N
Synonym C.I. 12140, 1-(2, 4-Dimethylphenylazo)-2-naphthol, Solvent orange 7
PubChem CID 5809936
IUPAC Name (1Z)-1-[(2,4-dimethylphenyl)hydrazinylidene]naphthalen-2-one
SMILES CC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(C)=C1

Specifications

Melting Point 156.0°C to 158.0°C
Infrared Spectrum Authentic
Lambda Max at approximately 420 and 493nm (in methanol)
Packaging Glass bottle
Specific Extinction 500 min. (at 493nm, in methanol)
Beilstein 16, 168
Color Brown/Orange to Red
Quantity 25 g
Physical Form Powder
Formula Weight 276.33
Grade Pure
Chemical Name or Material Sudan II
Show More Show Less
Hazard Category CANCER SUSPECTED AGENT
Hazard Statement GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.