Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan III 90.0+%, TCI America™
SDP

Catalog No. S0142100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0142100G 100 g
S014225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0142100G Supplier TCI America Supplier No. S0142100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-86-9
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.397
MDL Number MFCD00003905
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Synonym sudan iii, c.i. solvent red 23, 2-naphthalenol, 1-4-phenylazo phenyl azo, fhninjwbtrxebc-qszpnpogsa-n, sudaniii, sudan iiisolvent red 23, 2-naphthalenol, 1-2-4-2-phenyldiazenyl phenyl diazenyl, 1-2-4-2-phenyldiazen-1-yl phenyl diazen-1-yl naphthalen-2-ol, 1-e-4-e-phenylazo phenyl azo naphthalen-2-ol, 1-4-phenyldiazenylphenyl hydrazinylidene naphthalen-2-one
PubChem CID 6789251
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43

Specifications

Melting Point 202°C
Quantity 100 g
Formula Weight 352.40
Percent Purity ≥90.0% (E)
Physical Form Crystalline Powder
Chemical Name or Material Sudan III
EINECSNumber (5)-3087
RTECSNumber QK4250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.