Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sudan IV
SCI_ED

Catalog No. S25594 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25594 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25594
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 85-83-6
Molecular Formula C24H20N4O
Molecular Weight (g/mol) 380.45
MDL Number MFCD00003893
InChI Key KMDLOETUWUPGMB-BXCCFQQFSA-N
Synonym Oil Red, Fast Oil Red B
PubChem CID 6797604
IUPAC Name (1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
SMILES CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C

Specifications

Assay Percent Range 99.95% w/v
Linear Formula S
Formula Weight 380.17
Packaging Solid
Density 0.78g/mL
Quantity 10 g
Physical Form Powder
Chemical Name or Material Sudan IV
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.