accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNONNH3C(Z)(E/Z)

CAS RN 85-83-6

IUPAC Name: (1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
Synonyms: colouring agent CI 26105 (solvent red 24)
Molecular Weight (g/mol): 380.45
Molecular Formula: C24H20N4O
InChi Key: KMDLOETUWUPGMB-BXCCFQQFSA-N
SMILES: CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C
Molecular Weight (g/mol) 
  • (1)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
Grade 
  • (1)
  • (1)
  • (1)
  • (3)
Color 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (9)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)