Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan IV, MP Biomedicals™

Catalog No. ICN15264825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15264825 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15264825 Supplier MP Biomedicals, Inc Supplier No. 0215264825
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-83-6
Molecular Formula C24H20N4O
Molecular Weight (g/mol) 380.45
MDL Number MFCD00003893
InChI Key KMDLOETUWUPGMB-BXCCFQQFSA-N
Synonym Fat ponceau, Lipid crimson
PubChem CID 6797604
IUPAC Name 1-[[2-methyl-4-[(2-methylphenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-one
SMILES CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C

Specifications

Formula Weight 380.4
Color Brown to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Sudan IV
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.