Learn More
Invitrogen™ SUGP1 Polyclonal Antibody

Description
Immunogen sequence: MSLKMDNRDV AGKANRWFGV APPKSGKMNM NILHQEELIA QKKREIEAKM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Yeast: 83%; Zebrafish: 100%.
Specifications
Specifications
| Antigen | SUGP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SUGP1 |
| Gene Accession No. | Q8IWZ8 |
| Gene Alias | 5730496N02Rik; DKFZp434E2216; F23858; RBP; RNA-binding protein RBP; SF4; Splicing factor 4; SUGP1; SURP and G patch domain containing 1; SURP and G-patch domain containing 1; SURP and G-patch domain-containing protein 1 |
| Gene Symbols | SUGP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.