Learn More
Invitrogen™ SULF2 Polyclonal Antibody

Description
A suggested positive control for this product is human brain lysate.
Specifications
Specifications
| Antigen | SULF2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | SULF2 |
| Gene Accession No. | Q8IWU5 |
| Gene Alias | 2010004N24Rik; AU020235; DKFZp313E091; Extracellular sulfatase Sulf-2; Extracellular sulfatase Sulf-2-like protein; FLJ44488; FLJ90554; HSULF-2; KIAA1247; MGC126411; mKIAA1247; mSulf-2; OTTHUMP00000031223; RP5-1049G16.1; SULF2; sulfatase 2; sulfatase FP2; UNQ559/PRO1120 |
| Gene Symbols | SULF2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.