Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadiazine Sodium USP, Medisca™
GREENER_CHOICESDP

Catalog No. 18048055 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
25 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-055 25 kg
18-048-096 500 g
18-048-097 5 kg
18-048-098 100 g
18-048-115 2.5 kg
18-048-140 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-048-055 Supplier Medisca Supplier No. 322807
Add to Cart
Add to Cart

Sulfadiazine Sodium USP Not for Human Use.

Chemical Identifiers

CAS 547-32-0

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White
Quantity 25 kg
Percent Purity 99.0 to 100.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.