Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadimethoxine, ≥98.5% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111012798 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2798 10 g
11-101-2799 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2798 Supplier MilliporeSigma Supelco Supplier No. S700710G
Add to Cart
Add to Cart

Chemical structure: sulfonamide

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 122-11-2
Physical Form Neat
Linear Formula C12H14N4O4 S
MDL Number MFCD00057345
Synonym 4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide
Type Analytical Standard
Quantity 10 g
Formula Weight 310.33
Percent Purity ≥98.5% (TLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.