Learn More
Medchemexpress LLC HY-B0035 500mg Medchemexpress, Sulfamethazine CAS:57-68-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0035500MG
Encompass_Preferred
Medchemexpress, HY-B0035 500mg Sulfamethazine CAS:57-68-1 Sulfamethazine (Sulfadimidine) is an antimicrobial that is widely used to treat and prevent various animal diseases (such as gastrointestinal and respiratory tract infections). In China and the European Commission, the maximum residue level for Sulfamethazine in animal product is set at 100 µg/kg. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0095
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.