Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethoxazole, 100%, MP Biomedicals™

Catalog No. ICN15671125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15671125 25 g
ICN15671180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15671125 Supplier MP Biomedicals, Inc Supplier No. 0215671125
Add to Cart
Add to Cart

Chemical Identifiers

CAS 723-46-6
Molecular Formula C10H11N3O3S
Molecular Weight (g/mol) 253.276
InChI Key JLKIGFTWXXRPMT-UHFFFAOYSA-N
Synonym sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin
PubChem CID 5329
ChEBI CHEBI:9332
IUPAC Name 4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
SMILES CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Color White
Quantity 25 g
Formula Weight 253.3
Physical Form Powder
Chemical Name or Material Sulfamethoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.