Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, ACS Reagent, Reagents

Catalog No. 01337570
Change view
Click to view available options
Quantity:
500 g
6 lb.
50 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-570 6 lb.
01-337-572 500 g
01-337-571 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-570 Supplier Reagents Holdings Llc Supplier No. C21143006LB
Add to Cart
Add to Cart

Sulfamic Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.09
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid
IUPAC Name sulfamic acid
SMILES NS(O)(=O)=O

Specifications

Color White to Off-White
Melting Point 205°C
Packaging Poly Bottle
Specific Gravity 2.15
Quantity 6 lb.
Physical Form Solid
Grade ACS
Chemical Name or Material Sulfamic Acid, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.