Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, Analytical Standard (for Acidimetry), ACS reagent, 99.3-100.3% (dried material), MilliporeSigma™ Supelco™

Catalog No. 111018575 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8575 1 kg
11-102-0395 100 g
11-101-9032 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8575 Supplier MilliporeSigma Supelco Supplier No. 860401KG
Add to Cart
Add to Cart

Specifications

CAS 5329-14-6
Linear Formula NH2SO3H
MDL Number MFCD00011603
Technique Acidimetry
Melting Point 215°C to 225°C (decomposition)
Synonym Amidosulfonic acid
Quantity 1 kg
Formula Weight 97.09
Percent Purity 99.3 to 100.3% (dried material)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.