Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sulfanilamide 98%

Catalog No. AC132850025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC132850025 2.5 kg
AC132851000 100 g
AC132855000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC132850025 Supplier Thermo Scientific Chemicals Supplier No. 132850025
Add to Cart

Sulfanilamide, CAS # 63-74-1, is an organic sulfur compound that shows antibacterial activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sulfanilamide is an organic sulfur compound with antibacterial properties that are structurally similar to the p-aminobenzoic acid (PABA)
  • It also has a competitive inhibitory capacity against dihydropteroate synthetases blocking the synthesis of folic acid
  • Sulfanilamide competes with PABA for the bacterial enzyme dihydropteroate synthase preventing, the incorporation of PABA into dihydrofolic acid, an immediate precursor of folic acid. This culminates in the inhibition of the bacterial folic acid synthesis and also the de novo synthesis of purines and pyrimidines, resulting in cell growth arrest and cell death.

Applications

  • This compound shows antibacterial activity against different species, including gram-positive, gram-negative, and Chlamydia
  • In chemical reactions, this compound can act as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution
  • Sulfanilamide shows inhibitory activity against the enzyme, carbonic anhydrase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00007939
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym 4-Aminobenzenesulfonamide
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Type Sulfanilamide
Physical Form Crystalline Powder
Melting Point 163.0°C to 167.0°C
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (1 g, 105°C)
Packaging Plastic Bottle
Color White
Quantity 2.5 kg
Assay Percent Range 98%
Linear Formula 4-(H2N)C6H4SO2NH2
Beilstein 14, 698
Merck Index 15, 9057
Solubility Information Solubility in water: 7.5g/L (20°C).
Formula Weight 172.2
Percent Purity 98%
Chemical Name or Material Sulfanilamide, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-563-4
RTECSNumber WO8400000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.