Learn More
U.S. Pharmacopeia Sulfanilamide Melting Point Standard (Approximately 165 degrees), 63-74-1, MFCD00007939, 1g
Catalog No. NC9849080
Encompass
Molecular formula C6-H8-N2-O2-S, Molecular weight 172.21 g/mol, Melting Point 327.02 - 331.52 °F (163.9 - 166.4 °C), Synonyms: 4-Aminophenylsulfonamide, p-Aminobenzenesulfonamide
Catalog No. NC9849080 Supplier U.S. Pharmacopeia Supplier No. 1633007
Molecular formula C6-H8-N2-O2-S, Molecular weight 172.21 g/mol, Melting Point 327.02 - 331.52 °F (163.9 - 166.4 °C), Synonyms: 4-Aminophenylsulfonamide, p-Aminobenzenesulfonamide