Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sulfanilic Acid
SCI_ED

Catalog No. S25793A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25793A 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25793A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Specifications

CAS 121-57-3
Color White
Density 1.485g/mL
Assay Percent Range 100%
Packaging Solid
Molecular Formula C6H7NO3S
Linear Formula NH2C6H4SO3H·H2O
Quantity 100 g
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)O
IUPAC Name 4-aminobenzenesulfonic acid
Molecular Weight (g/mol) 173.186
PubChem CID 8479
ChEBI CHEBI:27500
Formula Weight 173.19
Chemical Name or Material Sulfanilic Acid
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.