Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilic Acid, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02004402 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-402 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-004-402 Supplier Avantor Supplier No. 035401
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Nitrogen Dioxide Determination (ASTM D-1607)

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
PubChem CID 8479
ChEBI CHEBI:27500
IUPAC Name 4-aminobenzenesulfonic acid
SMILES C1=CC(=CC=C1N)S(=O)(=O)O

Specifications

Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.49kg
Formula Weight 173.19
Percent Purity 99.8%
Grade BAKER ANALYZED™ ACS Reagent
Physical Form Powder
Chemical Name or Material Sulfanilic Acid, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.