Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilic acid sodium salt, MP Biomedicals™

Catalog No. ICN15671650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15671650 50 g
ICN15671683 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15671650 Supplier MP Biomedicals, Inc Supplier No. 0215671650
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6106-22-5
Molecular Formula C6H10NNaO5S
Molecular Weight (g/mol) 231.198
InChI Key MJFZIDUBUNIFLZ-UHFFFAOYSA-M
Synonym sodium sulfanilate dihydrate, unii-y31oen1192, 4-aminobenzenesulfonic acid, sodium salt dihydrate, sodium 4-aminobenzenesulfonate dihydrate, benzenesulfonic acid,4-amino-, sodium salt, hydrate 1:1:?, sulfanilic acid sodium salt dihydrate mi, acmc-1b9i8, c6h6nso3.na.2h2o, benzenesulfonic acid, 4-amino-, sodium salt, hydrate 1:1:2, 4h-1,4-benzoxazin-4-amine,2,3-dihydro hcl
PubChem CID 23687713
IUPAC Name sodium;4-aminobenzenesulfonate;dihydrate
SMILES C1=CC(=CC=C1N)S(=O)(=O)[O-].O.O.[Na+]

Specifications

Color White
Quantity 50 g
Formula Weight 195.2
Physical Form Powder
Chemical Name or Material Sulfanilic acid sodium salt
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.