Learn More
SPEX CERTIPREP LLC 3P SULFATE 500ML 1000PPM
Supplier:  SPEX CERTIPREP LLC 3P ICSSM500
Encompass_Preferred
(HPS by Zeptometrix) Sulfate (from Na2SO4) IC Std 1000 ug/mL in H2O Exp Date: 18 Months
Catalog No. NC9636386