Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfate Standard, 100 ppm SO42-, Ricca Chemical
SDP

Catalog No. R8110000120 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R8110000120 120 mL Natural Poly Bottle
8110-16 500 mL Natural Poly Bottle
811032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R8110000120 Supplier Ricca Chemical Company Supplier No. 81104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7757-82-6 , 7732-18-5
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.01
CAS Max % 100.0, 0.02
Equivalence 1 mL = 0.1 mg SO
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Sulfate Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.