Learn More
Sulfate Standard Stock Solution, 1000 ppm SO42-, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 98.0, 0.18 |
| CAS Max % | 100.0, 0.18 |
| pH | Approximately 7 |
| Boiling Point | 100°C |
| Quantity | 500 mL |
| Chemical Name or Material | SULFATE STANDARD STOCK SOLUTION |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.