Learn More
AdipoGen Sulfluramid
SDP
Supplier:  AdipoGen CDXS0050M250
Encompass_Preferred
Chemical. CAS 4151-50-2. Formula C10H6F17NO2S. MW 527.2. Synthetic. Perfluorinated delayed-action insecticide/pesticide. Mode of action is by uncoupling oxidative phosphorylation via disruption of the proton gradient. Suppresses immunoglobulin Ig M production in mice. Shown to be cytotoxic. Compound can be used as analytical reference material.
Catalog No. 50-256-0078
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.