Learn More
CUBE BIOTECH Nanodisc Copolymer Sulfo-DIBMA
Supplier:  CUBE BIOTECH 18533
Encompass_Preferred
10x1g of nanodisc copolymer Sulfo-DIBMA or the stabilization of membrane proteins outside the cell
Catalog No. NC2684575
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.