Learn More
Sigma Organic Chemistry Sulfo-LC-SDA (Sulfo-NHS-LC-Diazirine) (sulfosuccinimidyl 6-(4,4'-azipentanamido)hexanoate), | 50MG | MFCD28899432
Supplier: Sigma Organic Chemistry 80335950MG

Sulfo-LC-SDA (Sulfo-NHS-LC-Diazirine) (sulfosuccinimidyl 6-(4,4'-azipentanamido)hexanoate),, 50MG
About This Item:
Linear Formula: C15H21N4NaO8S
MDL Number: MFCD28899432
UNSPSC Code: 12352200
Storage: 2-8C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.