Learn More
Vector Laboratories Sulfo S-4FB Linker, 5 x 1 mg S-1011-010
SDP
Supplier:  Vector Laboratories S1011010
Encompass_Preferred
Sulfo S-4FB Linker (Water Soluble), 5 x 1 mg. S-4FB heterobifunctional crosslinker is fundamental to SoluLINK bioconjugation technology. Sulfo-S-4FB reacts with primary amines on proteins (through lysine amino acids) or amino-modified oligonucleotides or surfaces, introducing a 4FB linker that forms stable covalent conjugates with biomolecules possessing HyNic linkers. Sulfo-S-4FB contains a Sulfo-NHS ester, which makes this linker water soluble. No DMF or other organic solvents are necessary to dissolve the linker prior to use.
Catalog No. 50-204-5747
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.