Learn More
Thermo Scientific™ Sulfo-SBED Biotin Label Transfer Reagent, No-Weigh™ Format
Description
Thermo Scientific™ Pierce Sulfo-SBED Biotin Label Transfer Reagent is a multifunctional reagent for labeling a purified protein and then covalently transferring the attached biotin tag onto specific interactors of that protein.
Highlights:
- Sulfo-NHS ester reacts with primary amines at pH 7-9
- Aryl azide conjugates randomly when activated with UV light
- Cleavable disulfide bond allows biotin label transfer
- Discover protein interactions without radiolabeling
Includes:
No-Weigh Sulfo-SBED Label Transfer Reagent, 8 × 1mg; BupH Phosphate Buffered Saline, 1 Pk. (makes 500mL); Label Transfer Buffer (20X), 200mL (makes 4L); Streptavidin-HRP Conjugate, 0.1mg; No-Weigh Dithiothreitol (DTT), 8 × 7.7mg; Slide-A-Lyzer MINI Dialysis Units plus Float, 10K MWCO, 10-100μL, 10 units/Pk.
Specifications
Specifications
| Format | Powder |
| Quantity | 10 mg |
| Product Type | Crosslinker |
| Solubility | Water |
| Content And Storage | Upon receipt store at 4°C. |
| Labeling Method | Chemical Labeling, Label Transfer |
| Reactive Moiety | Sulfo-NHS Ester, Aryl Azide |
| Product Line | Pierce |
Frequently Asked Questions (FAQs)
Sulfo-SBED allows one to transfer a biotin tag from an amine-containing protein to a second protein (reacted with the phenyl azide). The amine-containing protein can be removed by reduction of the disulfide in the Sulfo-SBED spacer arm. This leaves just the second biotinylated protein for detection.
During the initial coupling procedure, the solutions should be protected from light to avoid decomposition of the phenyl azide functional group.
The NHS-ester end of the crosslinker is subject to hydrolysis, therefore the amine-containing protein should be conjugated prior to the photoactivatable (aryl azide) reaction.
UV light at wavelengths greater than 300 nm are appropriate for conjugation.
Yes, Sulfo-SBED is soluble in aqueous as well as organic solutions. It is most soluble in organics such as DMF and DMSO.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.