Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SulfoLink™ Immobilization Kit for Proteins, 2 mL

Catalog No. PI20325
Change view
Click to view available options
Quantity:
1 Column Kit
5 Column Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20325 1 Column Kit
PI44995 5 Column Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI20325 Supplier Thermo Scientific™ Supplier No. 20325
Add to Cart
Add to Cart

Covalently immobilize cysteine-peptides or proteins (sulfhydryl-ligands) for affinity purification using iodoacetyl-activated agarose beads and columns.

Thermo Scientific™ SulfoLink Coupling Resin is iodoacetyl-activated for the covalent immobilization of sulfhydryl-containing peptides or proteins for affinity purification.

SulfoLink Coupling Resin is porous, crosslinked 6% beaded agarose that has been activated with iodoacetyl groups. When incubated with a solution of peptide or protein that contains reduced cysteine residues, the iodoacetyl groups react specifically and efficiently with the exposed sulfhydryls (-SH) to form covalent and irreversible thioether bonds that permanently attach the peptide or protein to the resin. The result is a custom-made affinity resin for purification of antibodies, antigens and other molecules of interest.

Complete SulfoLink Immobilization Kits are available for peptides and proteins. Each kit contains all the reagents for preparing the respective peptide or protein sample and five columns containing 2mL of SulfoLink Resin that can be used in either gravity-flow or centrifuge format for efficient coupling reactions and multiple cycles of affinity purification. A single-column Trial Kit for use with either peptides or proteins is also available, or the SulfoLink Coupling Resin can be purchased separately.

Highlights:

  • Specific conjugation through sulfhydryl (-SH) groups – the iodoacetyl groups react specifically with sulhydryls to form irreversible thioether bonds

  • Separate kits optimized for peptides or proteins – kits include optimized reagents for preparing peptide or protein samples for efficient immobilization

  • Fast – spin columns increase protocol speed; prepare and couple samples in 2 hours (peptides) to 3.5 hours (proteins)

  • Flexible coupling conditions – use pH 7.5 to 9.0 aqueous buffers, organic solvent (e.g., 20% DMSO) or denaturant (guanidine·HCl), as needed for protein or peptide solubility during coupling reaction

  • Easy-to-follow instructions – streamlined protocols for sample preparation, immobilization and affinity purification

  • High capacity – immobilize 1 to 2mg peptide or 2 to 20mg protein per 2mL column of SulfoLink Coupling Resin

Includes:

Kits contain SulfoLink Resin Columns (2mL), protein or peptide preparation reagents (e.g., reducing agent), coupling buffer, wash solution, L-cysteine solution (for quenching) and desalting columns (protein kit only)

Recommended for:

Immobilize peptides having terminal cysteine residues to purify antibodies that were generated against peptide immunogens prepared by maleimide conjugation; Immobilize antibodies in oriented manner through hinge-region sulfhydryls to ensure that antigen binding sites are not sterically hindered for antigen affinity purification

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 mL
Content And Storage Sufficient For: Immobilizing 1 mg of peptide or 10 mg protein per column
• SulfoLink Column, 2 mL, 1 column
• SulfoLink Preparation Buffer, 7.5 mL
• SulfoLink Coupling Buffer, 120 mL
• Wash Solution, 25 mL
• PBS Pack (makes 500 mL), 1 pack
• 2-Mercaptoethylamine·HCl, 6 mg
• Bond-Breaker TCEP Solution, Neutral pH, 0.5 mL
• L-Cysteine·HCl, 100 mg
• Zeba Spin Desalting Column, 5 mL, 1 column
• Column Accessories

Upon receipt, store components at 4°C protected from light.
Product Line SulfoLink
Quantity 1 Column Kit
Type Immobilization Kit
Stationary Phase Haloacetyl
Sufficient For 1 to 20 mg of Peptide or Protein per Column
Format Spin Column
Column Type Affinity, Agarose Resin
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.