Learn More
Sulfonic Acid Polystyrene Resin cross-linked with 15% DVB (70-90mesh) (2.6-3.0mmol/g), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| Quantity | 5 g |
| MDL Number | MFCD02683442 |
| UN Number | 3261 |
| Synonym | Benzenesulfonic Acid Resin |
| Physical Form | Granules |
| Chemical Name or Material | Sulfonic Acid Polystyrene Resin cross-linked with 15% DVB (70-90mesh) (2.6-3.0mmol/g) |
Safety and Handling
| TSCA | No |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.