Learn More
Apexbio Technology LLC Congo Red 573-58-0 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B77665.1
Congo Red (CAS 573-58-0) is a small-molecule inhibitor targeting the vesicular glutamate transporter (VGlut) It is designed to modulate glutamate storage and release thereby influencing synaptic transmission Congo Red exerts its biological activity primarily through selective non-covalent interaction with -sheet-rich structures such as those found in amyloid fibrils Based on these properties Congo Red holds research potential in the detection and qualitative assessment of amyloid deposits in tissue sections characterization of amyloidosis-related samples plant and fungal cell wall amyloids and lipopolysaccharide staining in Gram-negative bacterial outer membranes for biomedical and microbiological studies
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.