Learn More
Abcam Sulforhodamine 101 Acid Chloride, Amine Reactive, 5 mg
Description
- Molecular Weight: 625.2 Da
- Purity: >95%
- Form: Powder
- Storage: Store at -20°C
- Shipping: Ambient - Can Ship with Ice
- Composition: 100% Sulforhodamine 101 acid chloride
- Description: Long wavelength red fluorescent amine-reactive rhodamine dye, also known as Texas Red. Useful for histological staining and cell sorting experiments with fluorescent-activated cell sorting machines (FACS).
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Label or Dye | Texas Red |
| For Use With (Equipment) | Fluorescent-activated Cell Sorting Machines |
| Buffer | 100% Sulforhodamine 101 acid chloride |
| Content And Storage | Store at -20°C. |
| Form | Powder |
| Detection Method | Fluorescence |
| Formulation | 100% Sulforhodamine 101 acid chloride |
| Quantity | 5 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.