Learn More
Medchemexpress LLC Sulforhodamine 101 DHPE | 187099-99-6 | >95.0% | 1381.84 | C74H117N4O14PS2 | 1MG
Supplier: Medchemexpress LLC HY1253851MG
Sulforhodamine 101 DHPE is a red fluorescent phospholipid probe created by conjugating dipalmitoyl phosphoethanolamine to sulforhodamine 101. It readily incorporates into lipid bilayers and is used for membrane imaging, monitoring probe colocalization in liposomes, and detecting protein-ligand interactions on supported bilayers. The dye exhibits excitation/emission peaks near 586/605 nm and is supplied as a powder with recommended cold storage.