Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfosalicylic Acid Solution (20%), Fisher Chemical

Catalog No. sa1681 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA168-1 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA168-1

Chemical Identifiers

CAS 97-05-2 , 7732-18-5
Molecular Formula C7H6O6S
Molecular Weight (g/mol) 218.179
MDL Number MFCD00007508
InChI Key YCPXWRQRBFJBPZ-UHFFFAOYSA-N
Synonym 5-sulfosalicylic acid, sulfosalicylic acid, 5-sulphosalicylic acid, salicylsulfonic acid, benzoic acid, 2-hydroxy-5-sulfo, salicylic acid, sulfo, benzoic acid, 2-hydroxysulfo, sulphosalicylic acid, salicylic acid, 5-sulfo, 2-hydroxybenzoic-5-sulfonic acid
PubChem CID 7322
ChEBI CHEBI:68555
IUPAC Name 2-hydroxy-5-sulfobenzoic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O

Specifications

Color Undesignated
Boiling Point 100°C
Assay Percent Range 19 to 21 %
Percent Purity 19 to 21%
Identification Pass Test
Packaging Glass Bottle
pH 0.25
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sulfosalicylic Acid Solution, 20%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.