Learn More
Chemplex Industries SULFUR IN DIESL FUEL STD 100ML
Supplier:  Chemplex Industries SDF200100
Encompass
For applications where there is little need to acquire complete sets of standards for monitoring the sulfur content in mineral, diesel or residual oil, single-element sulfur SpectroStandard Sulfur Standards are available. Each standard is formulated in its respective matrix with Di-N-butyl sulfide and accompanied with appropriate SpectroCertification of Analysis. Supplied in 100 cc quantities in glass bottles unless stated otherwise.
Catalog No. NC2542895
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.