Learn More
Sigma Organic Chemistry Sulfur monochloride | 250G | 10025-67-9 | MFCD00011446 | 0.98
Supplier: Sigma Organic Chemistry 157759250G

Sulfur monochloride, 250G
About This Item:
Synonym(s): Sulfur chloride
Linear Formula: S2Cl2
Molecular Weight: 135.04
CAS Number: 10025-67-9
MDL Number: MFCD00011446
UNSPSC Code: 12352302
Purity: 0.98
Boiling Point: 138 C (lit.)
Melting Point: -80 C (lit.)
Density: 1.688 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.658 (lit.)
Storage: room temp
EINECS Number: 233-036-2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.