Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™
SDP

Catalog No. p-4279733 Shop All Spex CertiPrep Products
Change view
Click to view available options
Concentration:
10 μg/g
100 μg/g
1000 μg/g
200 μg/g
25 μg/g
300 μg/g
5 μg/g
50 μg/g
75 μg/g
750 μg/g
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
DSS85Y 5 μg/g
DSS810Y 10 μg/g
DSS825Y 25 μg/g
DSS8AY 50 μg/g
DSS875Y 75 μg/g
DSS81Y 100 μg/g
DSS8BY 200 μg/g
DSS8CY 300 μg/g
DSS81BY 750 μg/g
DSS82Y 1000 μg/g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. DSS85Y Supplier SPEX CertiPrep Supplier No. DSS85Y
Only null left
Add to Cart
Add to Cart

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S5μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7704-34-9 , 8042-47-5
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.0, 99.99
CAS Max % 0.0, 99.99
Physical Form Solid
Concentration 5 μg/g
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Base Oil 20
Industry Type Petrochemical Testing
Quantity 100 mL
Chemical Name or Material Sulfur Oil Standard in Base Oil
Show More Show Less
DOTInformation Nonhazardous
Recommended Storage Ambient
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.