Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur Standard, 10,000μg/mL in water (low TOC, less-than 50 ppb), Ultra Scientific

Catalog No. USICP116
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICP116 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICP116 Supplier Agilent Technologies Supplier No. ICP116
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7704-34-9 , 7732-18-5
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Physical Form Liquid
Color Colorless
Quantity 125 mL
Chemical Name or Material Sulfur Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.